148202
4-Mercaptopyridine
95%
Synonym(s):
4-Pyridinethiol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H5NS
CAS Number:
Molecular Weight:
111.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
storage temp.
2-8°C
SMILES string
Sc1ccncc1
InChI
1S/C5H5NS/c7-5-1-3-6-4-2-5/h1-4H,(H,6,7)
InChI key
FHTDDANQIMVWKZ-UHFFFAOYSA-N
Related Categories
Application
4-Mercaptopyridine was used as a probe molecule to study the performance of the gold nanorod (AuNR)/polycaprolactone (PCL) fibrous mesh.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Peng Ji et al.
Frontiers in chemistry, 7, 245-245 (2019-05-07)
Direct observation of the surface-enhanced Raman scattering (SERS) of molecules adsorbed on nano-sized zirconia (ZrO2) substrates was first reported without the need for the addition of metal particles. It was found that ZrO2 nanoparticles can exhibit unprecedented Raman signal enhancements
Wenqiong Tang et al.
Analytical chemistry, 85(22), 10702-10709 (2013-10-22)
We report the fabrication of a homogeneous and highly dense gold nanorod (AuNR) assembly on electrospun polycaprolactone (PCL) fibers using electrostatic interaction as the driving force. Specifically, decoration of a poly(sodium 4-styrenesulfonate) (PSS) layer onto the AuNRs imposed negative charges
Jinglian Li et al.
ACS applied materials & interfaces, 3(10), 3936-3941 (2011-09-16)
A highly sensitive surface-enhanced Raman scattering (SERS) platform for the selective trace analysis of As(3+) ions was reported based on glutathione (GSH)/4-mercaptopyridine (4-MPY)-modified silver nanoparticles (AgNPs). Here, GSH conjugated on the surface of AgNPs for specifical binding with As(3+) ions
Yongmei Ma et al.
Nanomaterials (Basel, Switzerland), 9(3) (2019-03-16)
The high reproducibility of trace detection in complex systems is very hard but crucial to analytical technology and science. Here, we present a surface-enhanced Raman scattering (SERS) platform made by large-scale self-assembly of Au nanoparticle (NP) arrays at the cyclohexane/water
D Millo et al.
Langmuir : the ACS journal of surfaces and colloids, 23(8), 4340-4345 (2007-03-08)
To combine voltammetric techniques with surface-enhanced resonance Raman scattering (SERRS), cytochrome c (cyt c) was immobilized on a roughened silver electrode chemically modified with a self-assembled monolayer (SAM) of 4-mercaptopyridine (PySH). All measurements were performed on the same electrode in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service