Skip to Content
Merck
All Photos(1)

Key Documents

125881

Sigma-Aldrich

2,5-Dibromoaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2C6H3NH2
CAS Number:
Molecular Weight:
250.92
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

51-53 °C (lit.)

functional group

bromo

SMILES string

Nc1cc(Br)ccc1Br

InChI

1S/C6H5Br2N/c7-4-1-2-5(8)6(9)3-4/h1-3H,9H2

InChI key

WRTAZRGRFBCKBU-UHFFFAOYSA-N

Application

2,5-Dibromoaniline was used in the synthesis of 2,4,5-tribromoaniline. It was also used as starting reagent in the regioselective synthesis of 2-(3,4-dimethoxyphenyl)-5-bromobenzothiazole.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Induction of both cytochromes P-450 and P-448 by 2,3',4,4',5-pentabromobiphenyl, a component of fireMaster.
L W Robertson et al.
Biochemical and biophysical research communications, 92(1), 175-182 (1980-01-15)
Wei-Ming Liao et al.
Dalton transactions (Cambridge, England : 2003), 48(14), 4489-4494 (2019-03-13)
A new Eu-MOF was designed from an amino-functionalized ligand and Eu(iii) ions under solvothermal conditions. It is a highly porous, water-stable, and luminescent material, exhibiting pH sensing in the acidic range of pH = 7-3 with selective detection for Cd2+
Zong-Da Du et al.
Talanta, 206, 120179-120179 (2019-09-14)
In this work, the magnetic amino-functionalized microporous organic network composites (Fe3O4@MON-NH2) were rational designed and facile synthesized for magnetic solid phase extraction (MSPE) of endocrine disrupting chemicals (EDCs), followed by their analysis with high-performance liquid chromatography. The incorporation of amino
Du Zong-Da et al.
Journal of chromatography. A, 1616, 460791-460791 (2019-12-24)
Microporous organic network (MON) is a promise class of functional materials in solid phase extraction (SPE). However, the previous MON-based SPE works are all focused on off-line mode. The inherent drawbacks of off-line SPE such as complicated operation steps and
Catriona G Mortimer et al.
Journal of medicinal chemistry, 49(1), 179-185 (2006-01-06)
A series of new 2-phenylbenzothiazoles has been synthesized on the basis of the discovery of the potent and selective in vitro antitumor properties of 2-(3,4-dimethoxyphenyl)-5-fluorobenzothiazole (8n; GW 610, NSC 721648). Synthesis of analogues substituted in the benzothiazole ring was achieved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service