107565
1,1,1,3,3,3-Hexafluoro-2-phenyl-2-propanol
99%
Synonym(s):
α,α-Bis(trifluoromethyl)benzyl alcohol
About This Item
Recommended Products
Quality Level
Assay
99%
refractive index
n20/D 1.415 (lit.)
bp
160 °C (lit.)
density
1.45 g/mL at 25 °C (lit.)
functional group
fluoro
hydroxyl
phenyl
SMILES string
OC(c1ccccc1)(C(F)(F)F)C(F)(F)F
InChI
1S/C9H6F6O/c10-8(11,12)7(16,9(13,14)15)6-4-2-1-3-5-6/h1-5,16H
InChI key
IZPIZCAYJQCTNG-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
140.0 °F - closed cup
Flash Point(C)
60 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service