GC Analysis of Deprenyl (Selegiline) Enantiomers on Astec® CHIRALDEX™ B-DM
Materials
analytical column
Application
standard
CONDITIONS
column
Astec CHIRALDEX B-DM, 30 m x 0.25 mm I.D., 0.12 μm (77023AST)
oven
130 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 24 psi
sample
peak 1: R(-)-deprenyl (Selegiline)
peak 2: S(+)-deprenyl
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC