Merck
  • Subcritical water extraction combined with molecular imprinting technology for sample preparation in the detection of triazine herbicides.
Subcritical water extraction combined with molecular imprinting technology for sample preparation in the detection of triazine herbicides.
Journal of chromatography. A (2017-08-10)
Fengnian Zhao, Shanshan Wang, Yongxin She, Chao Zhang, Lufei Zheng, Maojun Jin, Hua Shao, Fen Jin, Xinwei Du, Jing Wang
ABSTRACT
A selective, environmentally friendly, and cost-effective sample extraction method based on a combination of subcritical water extraction (SWE) and molecularly imprinted solid-phase extraction (MISPE) was developed for the determination of eight triazine herbicides in soil samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS). In SWE, the highest extraction yields of triazine herbicides were obtained under 150°C for 15min using 20% ethanol as the organic modifier. Addition of MIP during SWE increased the extraction efficiency, and using MIP as a selective SPE sorbent improved the enrichment capability. Soil samples were treated with the optimized extraction MIP/SWE-MISPE method and analyzed by LC-MS/MS. The novel technique was then applied to soil samples for the determination of triazine herbicides, and better recoveries (78.9%-101%) were obtained compared with using SWE-MISPE (30%-67%). Moreover, this newly developed method displayed good linearity (R
Sigma-Aldrich
2-(Diethylamino)ethyl methacrylate, contains 1500 ppm MEHQ as inhibitor, 99%
Supelco
Simetryn, PESTANAL®, analytical standard
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
1-[2-(Trifluoromethyl)phenyl]imidazole