Merck
  • An HPLC method for the determination of selected amino acids in human embryo culture medium.
An HPLC method for the determination of selected amino acids in human embryo culture medium.
Biomedical chromatography : BMC (2016-07-28)
Petra Drábková, Lenka Andrlová, Roman Kanďár
ABSTRACT
A method for the determination of selected amino acids in culture medium using HPLC with fluorescence detection is described. Twenty hours after intra-cytoplasmic sperm injection, one randomly selected zygote was transferred to the culture medium. After incubation (72 h after fertilization), the culture medium in which the embryo was incubated and blank medium was immediately stored at -80°C. Filtered medium samples were derivatized with ortho-phthalaldehyde (naphthalene-2,3-dicarboxaldehyde), forming highly fluorescent amino acids derivatives. Reverse-phase columns (LichroCART, Purospher STAR RP
Product Description
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 300 μm
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 300 μm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 10 mm
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 500 μm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 1 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 7.5 cm × 4.6 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
Supelco
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 4.6 mm, pkg of 3 ea
Supelco
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 3 mm, pkg of 3 ea
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 500 μm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 3 cm × 4.6 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 7.5 cm × 3 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 10 cm × 3 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 3 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 3 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 3 cm × 3 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 3 cm × 2.1 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 7.5 cm × 2.1 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Ascentis® Express C18 PCP HPLC Columns, 2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® Express C18 PCP HPLC Columns, 2.7 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 7.5 cm × 3 mm
Supelco
Ascentis® Express C18, 5 μm Guard Cartridge, 5 μm particle size, L × I.D. 5 mm × 4.6 mm, pkg of 3 ea