Merck
  • The 1:2 complex between RavZ and LC3 reveals a mechanism for deconjugation of LC3 on the phagophore membrane.
The 1:2 complex between RavZ and LC3 reveals a mechanism for deconjugation of LC3 on the phagophore membrane.
Autophagy (2016-10-30)
Do Hoon Kwon, Sulhee Kim, Yang Ouk Jung, Kyung-Hye Roh, Leehyeon Kim, Byeong-Won Kim, Seung Beom Hong, In Young Lee, Ju Han Song, Woo Cheol Lee, Eui-Ju Choi, Kwang Yeon Hwang, Hyun Kyu Song
ABSTRACT
Hosts utilize macroautophagy/autophagy to clear invading bacteria; however, bacteria have also developed a specific mechanism to survive by manipulating the host cell autophagy mechanism. One pathogen, Legionella pneumophila, can hinder host cell autophagy by using the specific effector protein RavZ that cleaves phosphatidylethanolamine-conjugated LC3 on the phagophore membrane. However, the detailed molecular mechanisms associated with the function of RavZ have hitherto remained unclear. Here, we report on the biochemical characteristics of the RavZ-LC3 interaction, the solution structure of the 1:2 complex between RavZ and LC3, and crystal structures of RavZ showing different conformations of the active site loop without LC3. Based on our biochemical, structural, and cell-based analyses of RavZ and LC3, both distant flexible N- and C-terminal regions containing LC3-interacting region (LIR) motifs are important for substrate recognition. These results suggest a novel mechanism of RavZ action on the phagophore membrane and lay the groundwork for understanding how bacterial pathogens can survive autophagy.
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
Sigma-Aldrich
Polyethylenimine hydrochloride, linear, average Mn 10,000, PDI ≤1.5
Sigma-Aldrich
HRP-conjugated Anti-Rabbit IgG Concentrate (Item I1)