Merck
  • Structure and flexibility of the endosomal Vps34 complex reveals the basis of its function on membranes.
Structure and flexibility of the endosomal Vps34 complex reveals the basis of its function on membranes.
Science (New York, N.Y.) (2015-10-10)
Ksenia Rostislavleva, Nicolas Soler, Yohei Ohashi, Lufei Zhang, Els Pardon, John E Burke, Glenn R Masson, Chris Johnson, Jan Steyaert, Nicholas T Ktistakis, Roger L Williams
ABSTRACT
Phosphatidylinositol 3-kinase Vps34 complexes regulate intracellular membrane trafficking in endocytic sorting, cytokinesis, and autophagy. We present the 4.4 angstrom crystal structure of the 385-kilodalton endosomal complex II (PIK3C3-CII), consisting of Vps34, Vps15 (p150), Vps30/Atg6 (Beclin 1), and Vps38 (UVRAG). The subunits form a Y-shaped complex, centered on the Vps34 C2 domain. Vps34 and Vps15 intertwine in one arm, where the Vps15 kinase domain engages the Vps34 activation loop to regulate its activity. Vps30 and Vps38 form the other arm that brackets the Vps15/Vps34 heterodimer, suggesting a path for complex assembly. We used hydrogen-deuterium exchange mass spectrometry (HDX-MS) to reveal conformational changes accompanying membrane binding and identify a Vps30 loop that is critical for the ability of complex II to phosphorylate giant liposomes on which complex I is inactive.
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Avanti
Brain PE, Avanti Research - A Croda Brand 840022C
Avanti
Brain PE, Avanti Research - A Croda Brand 840022P, powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Avanti
Liver PI, Avanti Research - A Croda Brand
Avanti
Brain PC, Avanti Research - A Croda Brand 840053C
Avanti
Liver PI, Avanti Research - A Croda Brand
Avanti
Brain PS, Avanti Research - A Croda Brand
Avanti
Brain PC, Avanti Research - A Croda Brand 840053P, powder
Avanti
Brain PS, Avanti Research - A Croda Brand