Merck
  • Bioequivalence Study of Atorvastatin Tablets in Healthy Pakistani Volunteers.
Bioequivalence Study of Atorvastatin Tablets in Healthy Pakistani Volunteers.
Therapie (2015-02-14)
Sohail Mohammad, Usman Arshad, Nasir Abbass, Irfan Parvez, Ghulam Abbas, Wajahat Mahmood
ABSTRACT
A two way, randomized cross-over bioequivalence study was conducted to analyse the rate and extent of absorption of atorvastatin after a single dose of 80 mg atorvastatin as atorvastatin calcium tablets. The study was carried out using healthy male volunteers (N = 24). A high performance liquid chromatography method was employed to determine the level of drug in human plasma. It was concluded that the test and the reference drug exhibited comparable values of pharmacokinetic parameters. It was also concluded that since there was no significant difference between the rate and extent of absorption of the drug from the test and the reference formulations: these two formulations could thus be declared bioequivalent.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%