Merck
  • Automated production at the curie level of no-carrier-added 6-[(18)F]fluoro-L-dopa and 2-[(18)F]fluoro-L-tyrosine on a FASTlab synthesizer.
Automated production at the curie level of no-carrier-added 6-[(18)F]fluoro-L-dopa and 2-[(18)F]fluoro-L-tyrosine on a FASTlab synthesizer.
Journal of labelled compounds & radiopharmaceuticals (2015-05-27)
C Lemaire, L Libert, X Franci, J-L Genon, S Kuci, F Giacomelli, A Luxen
ABSTRACT
An efficient, fully automated, enantioselective multi-step synthesis of no-carrier-added (nca) 6-[(18)F]fluoro-L-dopa ([(18)F]FDOPA) and 2-[(18)F]fluoro-L-tyrosine ([(18)F]FTYR) on a GE FASTlab synthesizer in conjunction with an additional high- performance liquid chromatography (HPLC) purification has been developed. A PTC (phase-transfer catalyst) strategy was used to synthesize these two important radiopharmaceuticals. According to recent chemistry improvements, automation of the whole process was implemented in a commercially available GE FASTlab module, with slight hardware modification using single use cassettes and stand-alone HPLC. [(18)F]FDOPA and [(18)F]FTYR were produced in 36.3 ± 3.0% (n = 8) and 50.5 ± 2.7% (n = 10) FASTlab radiochemical yield (decay corrected). The automated radiosynthesis on the FASTlab module requires about 52 min. Total synthesis time including HPLC purification and formulation was about 62 min. Enantiomeric excesses for these two aromatic amino acids were always >95%, and the specific activity of was >740 GBq/µmol. This automated synthesis provides high amount of [(18)F]FDOPA and [(18)F]FTYR (>37 GBq end of synthesis (EOS)). The process, fully adaptable for reliable production across multiple PET sites, could be readily implemented into a clinical good manufacturing process (GMP) environment.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Potassium carbonate, meets USP testing specifications
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Hydriodic acid, 57 wt. %, distilled, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%