Merck
  • Liquid chromatographic method for toxic biogenic amines in foods using a chaotropic salt.
Liquid chromatographic method for toxic biogenic amines in foods using a chaotropic salt.
Journal of chromatography. A (2015-07-05)
Jian-Jun Zhong, Ningbo Liao, Tian Ding, Xingqian Ye, Dong-Hong Liu
ABSTRACT
Direct separation of biogenic amines by reversed-phase liquid chromatography (RPLC) is not an easy task because their basic and hydrophilic characteristics can lead to poor retention, column overloading, peak tailing, and hence low efficiency. Rather than routinely resorting to derivatization or using classical hydrophobic ion-pair reagents (IPR), this work proposes a new RPLC method making use of the chaotropic salt KPF6 as inorganic additive to an acidic acetonitrilic eluent to remedy the difficulties. Amine retention, overload behavior, peak shape, and column efficiency were significantly improved. The use of excess KPF6 led to a very slight decrease of amine retention. Depending on amine, the dependence of the logarithmic retention factor on the volume percent of acetonitrile could be reasonably linear or quite convex. Coupled with UV detection, the method was applied to trace analysis for six biogenic, aromatic or heterocyclic amines in three types of food after a sample cleanup, as necessary, by ion-pair extraction. The reliability of the whole analysis was demonstrated to be satisfactory. The proposed method outperforms existing methods in that it eliminates the need for long and cumbersome derivatization procedures without losing sensitivity; it also represents a good surrogate for classical ion-pair chromatography (IPC) because of the desirable hydrophilicity of chaotropic salts.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Tyramine, 98%, FG
Sigma-Aldrich
Tyramine, ≥98.0%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Tryptamine, ≥97%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
N-Benzylmethylamine, 97%
Sigma-Aldrich
Phenethylamine, 99%
Sigma-Aldrich
Phenethylamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Phenethylamine, ≥99%
Sigma-Aldrich
Histamine, ≥97.0%