Merck
  • Optimization of an analytical methodology for the simultaneous determination of different classes of ultraviolet filters in cosmetics by pressurized liquid extraction-gas chromatography tandem mass spectrometry.
Optimization of an analytical methodology for the simultaneous determination of different classes of ultraviolet filters in cosmetics by pressurized liquid extraction-gas chromatography tandem mass spectrometry.
Journal of chromatography. A (2015-06-21)
Marlene Vila, J Pablo Lamas, Carmen Garcia-Jares, Thierry Dagnac, Maria Llompart
ABSTRACT
A methodology based on pressurized liquid extraction (PLE) followed by gas chromatography-tandem mass spectrometry (GC-MS/MS) has been developed for the simultaneous analysis of different classes of UV filters including methoxycinnamates, benzophenones, salicylates, p-aminobenzoic acid derivatives, and others in cosmetic products. The extractions were carried out in 1mL extraction cells and the amount of sample extracted was only 100mg. The experimental conditions, including the acetylation of the PLE extracts to improve GC performance, were optimized by means of experimental design tools. The two main factors affecting the PLE procedure such as solvent type and extraction temperature were assessed. The use of a matrix matched approach consisting of the addition of 10μL of diluted commercial cosmetic oil avoided matrix effects. Good linearity (R(2)>0.9970), quantitative recoveries (>80% for most of compounds, excluding three banned benzophenones) and satisfactory precision (RSD<10% in most cases) were achieved under the optimal conditions. The validated methodology was successfully applied to the analysis of different types of cosmetic formulations including sunscreens, hair products, nail polish, and lipsticks, amongst others.
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzophenone, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Acetic anhydride, 99.5%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzophenone, purified by sublimation, ≥99%
Sigma-Aldrich
Acetic anhydride
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
(±)-Camphor, ≥95.5%
Sigma-Aldrich
2-Ethylhexyl 2-cyano-3,3-diphenylacrylate, 97%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC