Merck
  • LC-MS-based metabolite profiling of methanolic extracts from the medicinal and aromatic species Mentha pulegium and Origanum majorana.
LC-MS-based metabolite profiling of methanolic extracts from the medicinal and aromatic species Mentha pulegium and Origanum majorana.
Phytochemical analysis : PCA (2015-05-20)
Amani Taamalli, David Arráez-Román, Leila Abaza, Ihsan Iswaldi, Alberto Fernández-Gutiérrez, Mokhtar Zarrouk, Antonio Segura-Carretero
ABSTRACT
There has been increasing interest dedicated to the phenolic compounds with a view to their antioxidant and healthy properties. Recent studies have focused on plants from the Lamiaceae family with special interest in phenolic compounds antioxidant potential. The metabolite profile of methanolic extracts from two Lamiacea medicinal plants was investigated. Mentha pulegium and Origanum majorana methanolic extracts were analysed using reversed-phase ultra-high-performance liquid chromatography (RP-UHPLC) coupled to electrospray ionisation quadrupole time-of-flight mass spectrometry (ESI-QTOF-MS) detection in the negative ion mode. A total of 85 metabolites were characterised from different families, such as organic acids and derivatives, amino acids and derivatives, nucleosides, phenolic compounds as well as other polar metabolites, by using the MS and MS/MS information provided by the QTOF-MS. However, the total phenols and flavonoids were also quantified spectrophotometrically and they registered higher amounts in Mentha pulegium than in Origanum majorana extract. Gallocatechin was the major compound in M. pulegium extract whereas quercetin dimethyl ether, jaceidin and dihydrokaempferide were the major ones in O. majorana extract. The distribution of phenolic compounds in the methanolic extract showed a variation among studied plants. Mentha pulegium can be considered as a source of gallocatechin.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Sodium formate, 99.998% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis