Merck
  • Antioxidant activities and antiproliferative activity of Thai purple rice cooked by various methods on human colon cancer cells.
Antioxidant activities and antiproliferative activity of Thai purple rice cooked by various methods on human colon cancer cells.
Food chemistry (2015-06-05)
Rassarin Chatthongpisut, Steven J Schwartz, Jirawat Yongsawatdigul
ABSTRACT
The changes in anthocyanins, phenolic compounds, and antioxidant activities of Thai purple rice cooked by various cooking devices, including an electric rice cooker, an autoclave and a microwave oven, were investigated. In raw rice, cyanidin-3-glucoside (cy-3-glu) and peonidin-3-glucoside (pn-3-glu) are predominant anthocyanins, whereas protocatechuic acid (PCA) and vanillic acid (VA) are major free phenolic acids, and ferulic acid (FA) and VA are major bound phenolic acids. The microwave cooking method resulted in a marked loss of phenolics, anthocyanins and antioxidant activities (p<0.05). A decrease of cy-3-glu occurred concomitantly with an increase of PCA upon any cooking methods applied. A methanolic extract of rice cooked under sterilization exhibited the highest content of PCA and the highest inhibition of Caco-2 cell proliferation with an IC50 (16.11 μg/mL) comparable to that of raw rice. Thai purple rice cooked under sterilization could be a potential source of PCA exerting high antiproliferative activity.
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis