Merck
  • Protective effect of gallic acid against bleomycin-induced pulmonary fibrosis in rats.
Protective effect of gallic acid against bleomycin-induced pulmonary fibrosis in rats.
Pharmacological reports : PR (2015-10-21)
Jafar Nikbakht, Ali Asghar Hemmati, Ardeshir Arzi, Mohammad Taghi Mansouri, Anahita Rezaie, Mehri Ghafourian
ABSTRACT
Bleomycin (BLM), a chemotherapeutic agent is indicated in the management of some types of cancers. This drug produces a dose-dependent pulmonary fibrosis (PF) in most patients as well as experimental animals through oxidative injury. This study aimed to investigate the effect of gallic acid (GA), a polyphenolic compound, against PF-induce by BLM in rats. The rats were given GA orally at doses (50, 100, and 200 mg/kg/day) for 7 consecutive days before the administration of single intratracheal (it) instillation of BLM at 7.5 IU/kg. GA doses were continued for 21 days after BLM exposure. The regulatory effects of GA on BLM-induced pulmonary toxicity were determined by assaying oxidative stress biomarkers, lung and serum cytokine levels, and by histopathological examination of lung tissue. The results showed that intratracheal BLM administration significantly increased the inflammatory or fibrotic changes, collagen content, levels of malondialdehyde (MDA), and pro-inflammatory cytokines such as TNF-α and IL1β in lung. Also, it significantly decreased non-enzymatic (total thiol) and enzymatic (glutathione peroxidase (GPx)) antioxidant contents in the rats' lung tissue. However, oral administration of GA reversed all of these biochemical indices as well as histopathological alterations induced by BLM. Results of the present study demonstrate that GA, by its antioxidant properties, attenuates oxidative damage and fibrosis induced by BLM. Thus, an effective supplement with GA as an adjuvant therapy may be a very promising compound in reducing the side effects of BLM.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), Vetec, reagent grade, 98%
Sigma-Aldrich
Potassium chloride, Vetec, reagent grade, 99%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%