Skip to Content
Merck
All Photos(3)

Key Documents

V900068

Sigma-Aldrich

Potassium chloride

Vetec, reagent grade, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
reagent grade
form:
powder
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

99%

form

powder

pH

7

mp

770 °C (lit.)

density

1.98 g/mL at 25 °C (lit.)

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanyuan Qu et al.
Nucleic acids research, 41(10), 5263-5272 (2013-04-13)
Bacterial nucleoid-associated proteins, such as H-NS-like proteins in Enterobacteriaceae, are abundant DNA-binding proteins that function in chromosomal DNA organization and gene transcription regulation. The Mycobacterium tuberculosis Lsr2 protein has been proposed to be the first identified H-NS analogue in Gram-positive
Calista K L Ng et al.
Human molecular genetics, 24(11), 3163-3171 (2015-02-26)
mRNA decay is an essential and active process that allows cells to continuously adapt gene expression to internal and environmental cues. There are two mRNA degradation pathways: 3' to 5' and 5' to 3'. The DCPS protein is the scavenger
Masatoshi Takagaki et al.
Experimental neurology, 252, 12-17 (2013-11-20)
Sedatives in the neurointensive care unit can strongly influence patients' risks of developing secondary brain damage. In particular, isoflurane, a volatile anesthetic, has been recently re-introduced to the neurointensive care unit, and first clinical studies suggest beneficial effects due to
Hanne Sørup Tastesen et al.
Amino acids, 46(7), 1659-1671 (2014-03-25)
High-protein diets induce alterations in metabolism that may prevent diet-induced obesity. However, little is known as to whether different protein sources consumed at normal levels may affect diet-induced obesity and associated co-morbidities. We fed obesity-prone male C57BL/6J mice high-fat, high-sucrose
Linda K Månsson et al.
Faraday discussions, 181, 49-69 (2015-05-15)
We describe a route towards self-assembled colloidal molecules, where thermoresponsive microgels serve as discrete, externally tunable interaction sites. The ability of poly(N-isopropylacrylamide) (PNIPAM) and poly(N-isopropylmethacrylamide) (PNIPMAM) microgels to adsorb to the oil/water (O/W) interface and create Pickering-stabilized mini-emulsions was first

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service