Merck
  • Estimation of pressure-, temperature- and frictional heating-related effects on proteins' retention under ultra-high-pressure liquid chromatographic conditions.
Estimation of pressure-, temperature- and frictional heating-related effects on proteins' retention under ultra-high-pressure liquid chromatographic conditions.
Journal of chromatography. A (2015-04-01)
Szabolcs Fekete, Davy Guillarme
ABSTRACT
The goal of this work was to evaluate the changes in retention induced by frictional heating, pressure and temperature under ultra high pressure liquid chromatography (UHPLC) conditions, for four model proteins (i.e. lysozyme, myoglobin, fligrastim and interferon alpha-2A) possessing molecular weights between 14 and 20kDa. First of all, because the decrease of the molar volume upon adsorption onto a hydrophobic surface was more pronounced for large molecules such as proteins, the impact of pressure appears to overcome the frictional heating effects. Nevertheless, we have also demonstrated that the retention decrease due to frictional heating was not negligible with such large biomolecules in the variable inlet pressure mode. Secondly, it is clearly shown that the modification of retention under various pressure and temperature conditions cannot be explained solely by the frictional heating and pressure effects. Indeed, some very uncommon van't Hoff plots (concave plots with a maximum) were recorded for our model/therapeutic proteins. These maximum retention factors values on the van't Hoff plots indicate a probable change of secondary structure/conformation with pressure and temperature. Based on these observations, it seems that the combination of pressure and temperature causes the protein denaturation and this folding-unfolding procedure is clearly protein dependent.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent