Merck
  • Imprinted nanospheres based on precipitation polymerization for the simultaneous extraction of six urinary benzene metabolites from urine followed by injector port silylation and gas chromatography-tandem mass spectrometric analysis.
Imprinted nanospheres based on precipitation polymerization for the simultaneous extraction of six urinary benzene metabolites from urine followed by injector port silylation and gas chromatography-tandem mass spectrometric analysis.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-08-11)
Abhishek Chauhan, Tejasvi Bhatia, Manoj Kumar Gupta, Pathya Pandey, Vivek Pandey, Prem Narain Saxena, Mohana Krishna Reddy Mudiam
ABSTRACT
In the present communication, uniformly sized molecularly imprinted polymer (MIP) as nanospheres were synthesized based on precipitation polymerization using dual-template imprinting approach and used it as sorbent for solid phase extraction of six urinary benzene metabolites (UBMs). This approach in combination with injector port silylation (IPS) has been used for the quantitative determination of these UBMs by gas chromatography-tandem mass spectrometry. The MIP was synthesized by using t,t-muconic acid (t,t-MA) and 1,2,4-trihydroxybenzene (THB) as templates, methacrylic acid (MAA) as a monomer, ethyleneglycoldimethacrylate (EGDMA) as crosslinker, acetonitrile and dimethylsulphoxide as a porogen and azobisisobutyronitrile (AIBN) as an initiator. The factors affecting the performance of polymer and IPS were investigated and optimized for the simultaneous determination of UBMs in urine. Binding study of imprinted and non-imprinted polymer (NIP) shows that, MIP possesses higher affinity in comparison to NIP for these analytes. Under the optimum conditions, the method developed was found to be linear with regression coefficients falls in the range of 0.9721-0.9988 for all the analyzed metabolites. The percent recovery of the metabolites analyzed in urine was found to be in the range of 76-89%, while the limit of detection and limit of quantification were found to be in the range of 0.9-9.1ngmL(-1) and 2.8-27ngmL(-1) respectively. The validated method was successfully applied to the real urine samples collected from different groups (kitchen workers, smokers and petroleum workers) and found that the developed method has been promising applications in the routine analysis of urine samples of benzene exposed population.
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Pyrocatechol, ≥99%
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Sigma-Aldrich
1-[2-(Trifluoromethyl)phenyl]imidazole
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Divinylbenzene, technical grade, 55%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Divinylbenzene, technical grade, 80%
Sigma-Aldrich
Trifluoroacetamide, 97%
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%
Sigma-Aldrich
4-Vinylpyridine, contains 100 ppm hydroquinone as inhibitor, 95%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%