Merck
  • Development and Effects of FTY720 Ophthalmic Solution on Corneal Allograft Survival.
Development and Effects of FTY720 Ophthalmic Solution on Corneal Allograft Survival.
Scientific reports (2015-11-13)
Zhaochuan Liu, Haotian Lin, Chulong Huang, Wan Chen, Wu Xiang, Yu Geng, Weirong Chen
ABSTRACT
Fingolimod (FTY720), a novel class of sphingosine 1-phosphate receptor modulators, has received special interest among ophthalmologists, particularly given that oral administration of FTY720 has proven to effectively treat corneal graft rejection in animal models. However, no studies have examined the performance of FTY720 as an ophthalmic solution in reducing corneal rejection in high-risk corneal rejection models, and the stability and ocular irritation profile of FTY720 ophthalmic solution are also unknown. Thus, we developed 0.1%, 0.2% and 0.5% FTY720 ophthalmic solutions and evaluated their chemical stabilities under various storage conditions with high- performance liquid chromatography. To investigate the ocular irritancy of the FTY720 ophthalmic solution, New Zealand albino rabbits were subjected to the Draize test. Furthermore, classic, well-established rat allogenic penetrating keratoplasty models were used to investigate the anti-rejection efficacy of the tested FTY720 ophthalmic solutions. We found that the non-irritating 0.5% FTY720 ophthalmic solution could prolong corneal allograft survival in rats with significant efficacy for about one month. Furthermore, no significant concentration changes occurred in any of the types of FTY720 ophthalmic solutions within three months. These results revealed crucial profiles of FTY720 ophthalmic solutions and warrant further investigation and optimization of FTY720 in the anti-rejection therapy after keratoplasty.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%