Merck
  • Inflammatory bowel disease associates with proinflammatory potential of the immunoglobulin G glycome.
Inflammatory bowel disease associates with proinflammatory potential of the immunoglobulin G glycome.
Inflammatory bowel diseases (2015-04-22)
Irena Trbojević Akmačić, Nicholas T Ventham, Evropi Theodoratou, Frano Vučković, Nicholas A Kennedy, Jasminka Krištić, Elaine R Nimmo, Rahul Kalla, Hazel Drummond, Jerko Štambuk, Malcolm G Dunlop, Mislav Novokmet, Yurii Aulchenko, Olga Gornik, Harry Campbell, Maja Pučić Baković, Jack Satsangi, Gordan Lauc
ABSTRACT
Glycobiology is an underexplored research area in inflammatory bowel disease (IBD), and glycans are relevant to many etiological mechanisms described in IBD. Alterations in N-glycans attached to the immunoglobulin G (IgG) Fc fragment can affect molecular structure and immunological function. Recent genome-wide association studies reveal pleiotropy between IBD and IgG glycosylation. This study aims to explore IgG glycan changes in ulcerative colitis (UC) and Crohn's disease (CD). IgG glycome composition in patients with UC (n = 507), CD (n = 287), and controls (n = 320) was analyzed by ultra performance liquid chromatography. Statistically significant differences in IgG glycome composition between patients with UC or CD, compared with controls, were observed. Both UC and CD were associated with significantly decreased IgG galactosylation (digalactosylation, UC: odds ratio [OR] = 0.71; 95% confidence interval [CI], 0.5-0.9; P = 0.01; CD: OR = 0.41; CI, 0.3-0.6; P = 1.4 × 10) and significant decrease in the proportion of sialylated structures in CD (OR = 0.46, CI, 0.3-0.6, P = 8.4 × 10). Logistic regression models incorporating measured IgG glycan traits were able to distinguish UC and CD from controls (UC: P = 2.13 × 10 and CD: P = 2.20 × 10), with receiver-operator characteristic curves demonstrating better performance of the CD model (area under curve [AUC] = 0.77) over the UC model (AUC = 0.72) (P = 0.026). The ratio of the presence to absence of bisecting GlcNAc in monogalactosylated structures was increased in patients with UC undergoing colectomy compared with no colectomy (FDR-adjusted, P = 0.05). The observed differences indicate significantly increased inflammatory potential of IgG in IBD. Changes in IgG glycosylation may contribute to IBD pathogenesis and could alter monoclonal antibody therapeutic efficacy. IgG glycan profiles have translational potential as IBD biomarkers.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Anthranilamide, ≥98%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture