Merck
  • Continuous hydrolysis of carboxymethyl cellulose with cellulase aggregates trapped inside membranes.
Continuous hydrolysis of carboxymethyl cellulose with cellulase aggregates trapped inside membranes.
Enzyme and microbial technology (2015-07-29)
Le Truc Nguyen, Kristyn Rui Shan Neo, Kun-Lin Yang
ABSTRACT
Enzymatic hydrolysis of cellulose is often conducted in batch processes in which hydrolytic products tend to inhibit enzyme activity. In this study, we report a method for continuous hydrolysis of carboxymethyl cellulose (CMC) by using cross-linked cellulase aggregate (XCA) trapped inside a membrane. XCA particles prepared by using a millifluidic reactor have a uniform size distribution around 350 nm. Because of their large size, XCA particles in solutions can be filtered through a polyethersulfone membrane to collect 87.1 ± 0.9% of XCA particles. The membrane with impregnated XCA can be used as a catalyst for hydrolysis of CMC in a continuous mode. When the CMC concentration is 1.0 g/l and the flow rate is 2 μl/min, 53.9% of CMC is hydrolyzed to reducing sugars. The membrane with XCA is very stable under continuously flowing solutions. After 72 h of reaction, 97.5% of XCA remains inside the membrane.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
3,5-Dinitrosalicylic acid, used in colorimetric determination of reducing sugars
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
3,5-Dinitrosalicylic acid, 98%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, Vetec, reagent grade, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis