Merck
Dry powder formulation of simvastatin.
Expert opinion on drug delivery (2014-09-23)
Alaa S Tulbah, Hui Xin Ong, Lucy Morgan, Paolo Colombo, Paul M Young, Daniela Traini
ABSTRACT
This study focuses on the development of a dry powder inhaler (DPI) formulation of simvastatin (SV), and the effects of SV on the respiratory epithelium. Micronised SV samples were prepared by dry jet-milling. The long-term chemical stability and physicochemical properties of the formulations were characterised in terms of particles size, morphology, thermal and moisture responses. Furthermore, in vitro aerosol depositions were performed. The formulation was evaluated for cell viability and its effect on cilia beat activity, using ciliated nasal epithelial cells in vitro. The formulation transport across an established air interface Calu-3 bronchial epithelial cells and its ability to reduce mucus secretion was also investigated. The particle size of the SV formulation and its aerosol performance were appropriate for inhalation therapy. Moreover, the formulation was found to be non-toxic to pulmonary epithelia cells and cilia beat activity up to a concentration of 10(-6) M. Transport studies revealed that SV has the ability to penetrate into airway epithelial cells and is converted into its active SV hydroxy acid metabolite. Single dose of SV DPI also decreased mucus production after 4 days of dosing. This therapy could potentially be used for the local treatment of diseases like chronic obstructive pulmonary disease, cystic fibrosis, and bronchiectasis given its anti-inflammatory effects and ability to reduce mucus production.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium bromide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium bromide, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Sodium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SAFC
L-Glutamine
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%