Merck
  • Comparing immunocompetent and immunodeficient mice as animal models for bone tissue engineering.
Comparing immunocompetent and immunodeficient mice as animal models for bone tissue engineering.
Oral diseases (2015-02-05)
Y Zhang, X Li, T Chihara, T Mizoguchi, A Hori, N Udagawa, H Nakamura, H Hasegawa, A Taguchi, A Shinohara, H Kagami
ABSTRACT
To understand the differences and similarities between immunocompetent and immunodeficient mice as ectopic transplantation animal models for bone tissue engineering. Osteogenic cells from mouse leg bones were cultured, seeded on β-TCP granules, and transplanted onto the backs of either immunocompetent or immunodeficient nude mice. At 1, 2, 4, and 8 weeks postoperatively, samples were harvested and evaluated by hematoxylin-eosin staining, tartrate-resistant acid phosphatase (TRAP) staining, and immunohistochemical staining and quantitative PCR. In immunocompetent mice, inflammatory cell infiltration was evident at 1 week postoperatively and relatively higher expression of TNF-α and IL-4 was observed. In immunodeficient mice, new bone area and the number of TRAP-positive cells were larger at 4 weeks than in immunocompetent mice. The volume of new bone area in immunodeficient mice was reduced by 8 weeks. Bone regeneration was feasible in immunocompetent mice. However, some differences were observed between immunocompetent and immunodeficient mice in the bone regeneration process possibly due to different cytokine expression, which should be considered when utilizing in vivo animal models.
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
1-Naphthol, puriss. p.a., reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α from mouse, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Citric acid trisodium salt, ≥98% (GC), anhydrous
Sigma-Aldrich
Trisodium citrate dihydrate, meets USP testing specifications
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
1-Naphthol, BioXtra, ≥99%
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sp7
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture