Merck
  • Oxidative Stress Increases Surface Toll-Like Receptor 4 Expression in Murine Macrophages Via Ceramide Generation.
Oxidative Stress Increases Surface Toll-Like Receptor 4 Expression in Murine Macrophages Via Ceramide Generation.
Shock (Augusta, Ga.) (2015-05-07)
Patrick S Tawadros, Kinga A Powers, Menachem Ailenberg, Simone E Birch, John C Marshall, Katalin Szaszi, Andras Kapus, Ori D Rotstein
ABSTRACT
Multiorgan failure is a major cause of late mortality following trauma. Oxidative stress generated during shock/resuscitation contributes to tissue injury by priming the immune system for an exaggerated response to subsequent inflammatory stimuli, such as lipopolysaccharide (LPS). We recently reported that oxidative stress causes rapid recruitment of the LPS receptor Toll-like receptor 4 (TLR4) to membrane lipid rafts, thus increasing LPS responsiveness and cellular priming. We hypothesized that activation of Src family kinases by oxidants might contribute to these events. We utilized microscopy, flow cytometry, Western blotting, and thin-layer chromatography methods. Using hydrogen peroxide in vitro and hemorrhagic shock/resuscitation in vivo, oxidant-induced TLR4 translocation in macrophages occurred in an Src-dependent manner. Approaches supporting this conclusion included pharmacologic inhibition of the Src family kinases by PP2, Src inhibition by a molecular approach of cell transfection with Csk, and genetic inhibition of all Src kinases relevant to the monocyte/macrophage lineage in hckfgrlyn triple knockout mice. To evaluate the upstream molecules involved in Src activation, we evaluated the ability of oxidative stress to activate the bioactive lipid molecule ceramide. Oxidants induced ceramide generation in macrophages both in vitro and in vivo, an effect that appears to be due to activation of the acid sphingomyelinase. Using pharmacological approaches, ceramide was shown to be both necessary and sufficient to mediate TLR4 translocation to the plasma membrane in an Src-dependent manner. This study identifies a hierarchy of signaling molecules following oxidative stress that might represent novel targets for therapy in critical illness and organ injury.
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Lithium chloride, Vetec, reagent grade
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
G 418 disulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
G 418 disulfate salt, suitable for plant cell culture, BioReagent
Sigma-Aldrich
O-Tricyclo[5.2.1.02,6]dec-9-yl dithiocarbonate potassium salt, ≥95%, solid
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
SAFC
Glycine
Sigma-Aldrich
G 418 disulfate salt, potency: ≥720 μg per mg (dried basis)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O