Merck
  • Long-circulating lipoprotein-mimic nanoparticles for smart intravenous delivery of a practically-insoluble antineoplastic drug: Development, preliminary safety evaluations and preclinical pharmacokinetic studies.
Long-circulating lipoprotein-mimic nanoparticles for smart intravenous delivery of a practically-insoluble antineoplastic drug: Development, preliminary safety evaluations and preclinical pharmacokinetic studies.
International journal of pharmaceutics (2015-08-09)
Mina Ibrahim Tadros, Abdulaziz Mohsen Al-Mahallawi
ABSTRACT
Chlorambucil (CHL) is a water-insoluble antineoplastic drug having a short elimination half-life. It suffers from remarkable differences in pharmacokinetics following oral administration. The current work aimed to assess safety and pharmacokinetics of CHL-loaded, lipoprotein-mimic, nanoparticles (NPs) following intravenous administration. The design of NPs was based on complexation between egg yolk lecithin (EYL) and bovine serum albumin (BSA). The NPs were preliminary evaluated via FT-IR, DSC and P-XRD. The NPs were characterized for particle size, zeta potential, morphology and drug entrapment efficiency (EE%). The best achieved NP dispersion (LP6) and CHL solution were challenged for in vitro hemolytic potential, in vivo vascular irritation studies in rabbits and in vivo pharmacokinetics following intravenous administration in rats. The results confirmed that NPs were stabilized by hydrophobic-attractions and hydrogen-bondings between CHL, BSA and EYL. The amorphous dispersion of CHL within NPs was revealed. LP6 dispersion displayed monodispersed nano-spherical particles (144.33 ± 2.17 nm). It possessed the highest negative zeta potential (-30.55 ± 0.24 mV) and the largest EE% (86.35 ± 2.33%). The significantly (P < 0.05) prolonged MRT(0-∞), longer elimination t50% and reduced plasma clearance highlighted the long-circulating characteristics of LP6. The preliminary safety evaluations and the seven-fold increase in bioavailability elucidated potentiality for smart intravenous delivery of CHL.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Chlorambucil
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, meets USP testing specifications