Merck
  • Use of LC-MS/TOF, LC-MS(n), NMR and LC-NMR in characterization of stress degradation products: Application to cilazapril.
Use of LC-MS/TOF, LC-MS(n), NMR and LC-NMR in characterization of stress degradation products: Application to cilazapril.
Journal of pharmaceutical and biomedical analysis (2015-04-19)
Mallikarjun Narayanam, Archana Sahu, Saranjit Singh
ABSTRACT
Forced degradation studies on cilazapril were carried out according to ICH and WHO guidelines. Significant degradation of the drug was observed in acid and base conditions, resulting primarily in cilazaprilat. In neutral condition, five degradation products were formed, while under oxidative condition, two degradation products were generated. In total, seven degradation products were formed, which were separated on an Inertsil C-18 column using a stability-indicating HPLC method. Structure elucidation of the degradation products was done by using sophisticated and hyphenated tools like, LC-MS/TOF, LC-MS(n), on-line H/D exchange, LC-NMR and NMR. Initially, comprehensive mass fragmentation pathway of the drug was laid down. Critical comparison of mass fragmentation pathways of the drug and its hydrolytic degradation products allowed structure characterization of the latter. 1D and 2D proton LC-NMR studies further confirmed the proposed structures of hydrolytic degradation products. The oxidative degradation products could not be characterized using LC-MS and LC-NMR tools. Hence, these degradation products were isolated using preparative HPLC and extensive 1D ((1)H, (13)C, DEPT) and 2D (COSY, TOCSY, HETCOR and HMBC) NMR studies were performed to ascertain their structures. Finally, degradation pathways and mechanisms of degradation of the drug were outlined.
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis