Merck
  • MicroRNA-183 increases osteoclastogenesis by repressing heme oxygenase-1.
MicroRNA-183 increases osteoclastogenesis by repressing heme oxygenase-1.
Bone (2015-07-15)
Ke Ke, Ok-Joo Sul, Monisha Rajasekaran, Hye-Seon Choi
ABSTRACT
Emerging evidence suggests that microRNAs (miRs) influence skeletal structure by modulating osteoclastogenesis and bone resorption. We have demonstrated previously that the up-regulation of heme oxygenase-1 (HO-1) attenuated osteoclastogenesis in bone marrow-derived macrophages (BMMs). RANKL-induced osteoclastogenesis elevates microRNA-183 (miR-183) in BMM. We show here that HO-1 is a target gene of miR-183 and that this miRNA binds to the 3'-UTR of HO-1. We find that a synthetic inhibitor that binds to miR-183 decreases osteoclast (OC) differentiation and increases the expression of HO-1, while a mimic of endogenous mature miR-183 has the opposite effect. Moreover, the HO-1 inducers, resveratrol and piceatannol decrease the expression of miR-183, resulting in attenuated osteoclastogenesis. Our findings reveal how miR-183 affects OC formation.
Sigma-Aldrich
Piceatannol, powder
Sigma-Aldrich
MISSION® esiRNA, targeting human HMOX1
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Hmox1
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a. plus, ≥25% NH3 in H2O
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a., reag. ISO, reag. Ph. Eur., ~25% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium hydroxide solution, puriss., 30-33% NH3 in H2O
Sigma-Aldrich
Ammonium hydroxide solution, puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis
Sigma-Aldrich
Resveratrol, Vetec, reagent grade, 98%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%