Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Pricing and availability is not currently available.
Pricing and availability is not currently available.
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder or crystals
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Pricing and availability is not currently available.
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Pricing and availability is not currently available.
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, 99%, FCC
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium acetate solution, BioUltra, for molecular biology, 5 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Octylphosphonic acid, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Phthaldialdehyde, for fluorescence, ≥99.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamic acid, FCC
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium acetate, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium acetate, BioUltra, for molecular biology, ≥99.0% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium acetate, 99.98% trace metals basis
Pricing and availability is not currently available.