Merck
  • Chemopreventive and therapeutic potentials of thymoquinone in HepG2 cells: mechanistic perspectives.
Chemopreventive and therapeutic potentials of thymoquinone in HepG2 cells: mechanistic perspectives.
Journal of natural medicines (2015-03-23)
Abeer ElKhoely, Hafez F Hafez, Abeer M Ashmawy, Osama Badary, Ahmed Abdelaziz, Adel Mostafa, Samia A Shouman
ABSTRACT
Liver cancer is the fifth commonest malignancy worldwide and the third leading cause of death. Identifying novel curative and preventive therapy may improve its prognosis. In this study, thymoquinone (TQ), the most active biological ingredient of Nigella sativa Linn, was investigated for its antitumor activity. Mechanistic perspectives underlying this antitumor activity were explored by testing its effect on cell cycle, apoptosis, and angiogenesis. In addition, the chemopreventive effect of TQ was carried out by measuring its effect on phase I CYP1A1 and phase II glutathione S-transferase (GST) drug-metabolizing enzymes. The results of the present study revealed the effectiveness of TQ as an antitumor agent against different types of cancer including brain, colon, cervix and liver at both a time- and concentration-dependent manner. In HepG2 cells, it induced G2/M phase cell cycle arrest and a concentration-dependent increase in the percentage of apoptotic cells with an increase in the ratio of Bax/BCL-2. Moreover, the expression of mRNA and protein level of vascular endothelial growth factor decreased as the concentration of TQ increased. Our data showed a significant inhibition of induced phase I CYP1A1 enzyme, and elevation in the content of glutathione and activity of phase II enzyme GST, in HepG2 cells. Our results provide support for the beneficial use of TQ as a therapeutic and chemopreventive agent against liver cancer.
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, 97%
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
Thymoquinone, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
MISSION® esiRNA, targeting human BAX
Sigma-Aldrich
MISSION® esiRNA, targeting human NKX3-1
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
L-Glutathione reduced, Vetec, reagent grade, ≥98%