Merck
  • Cholesterol oxidase with high catalytic activity from Pseudomonas aeruginosa: Screening, molecular genetic analysis, expression and characterization.
Cholesterol oxidase with high catalytic activity from Pseudomonas aeruginosa: Screening, molecular genetic analysis, expression and characterization.
Journal of bioscience and bioengineering (2015-01-13)
Noriyuki Doukyu, Shyou Nihei
ABSTRACT
An extracellular cholesterol oxidase producer, Pseudomonas aeruginosa strain PA157, was isolated by a screening method to detect 6β-hydroperoxycholest-4-en-3-one-forming cholesterol oxidase. On the basis of a putative cholesterol oxidase gene sequence in the genome sequence data of P. aeruginosa strain PAO1, the cholesterol oxidase gene from strain PA157 was cloned. The mature form of the enzyme was overexpressed in Escherichia coli cells. The overexpressed enzyme formed inclusion bodies in recombinant E. coli cells grown at 20 °C and 30 °C. A soluble and active PA157 enzyme was obtained when the recombinant cells were grown at 10 °C. The purified enzyme was stable at pH 5.5 to 10 and was most active at pH 7.5-8.0, showing optimal activity at pH 7.0 and 70 °C. The enzyme retained about 90% of its activity after incubation for 30 min at 70 °C. The enzyme oxidized 3β-hydroxysteroids such as cholesterol, β-cholestanol, and β-sitosterol at high rates. The Km value and Vmax value for the cholesterol were 92.6 μM and 15.9 μmol/min/mg of protein, respectively. The Vmax value of the enzyme was higher than those of commercially available cholesterol oxidases. This is the first report to characterize a cholesterol oxidase from P. aeruginosa.
Product Description
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
N-Lauroylsarcosine sodium salt solution, 30% aqueous solution, ≥97.0% (HPLC)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
N-Lauroylsarcosine sodium salt solution, 20%, for molecular biology
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Phenol solution, BioReagent, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, ≥97.0% (HPLC)
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, BioUltra, for molecular biology, ≥99.0% (HPLC)
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
4-Aminoantipyrine, puriss. p.a., reag. Ph. Eur., ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
N-Lauroylsarcosine sodium salt, BioXtra, ≥97% (TLC)
Sigma-Aldrich
4-Aminoantipyrine, reagent grade
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer