Merck
  • Resveratrol attenuates CXCL11 expression induced by proinflammatory cytokines in retinal pigment epithelial cells.
Resveratrol attenuates CXCL11 expression induced by proinflammatory cytokines in retinal pigment epithelial cells.
Cytokine (2015-04-22)
R Krishnan Kutty, William Samuel, Rebecca Abay, Aswini Cherukuri, Chandrasekharam N Nagineni, Todd Duncan, Cynthia Jaworski, Camasamudram Vijayasarathy, T Michael Redmond
ABSTRACT
Dysfunction of the retinal pigment epithelium (RPE) resulting from chronic inflammation is implicated in the pathogenesis of age-related macular degeneration (AMD). RPE cells adjacent to drusen deposits in the AMD eye are known to contain CXCL11, a chemokine involved in inflammatory cell recruitment. We investigated the CXCL11 production by the human RPE (ARPE-19) cells under inflammatory conditions and tested its response to resveratrol, a naturally occurring anti-inflammatory antioxidant. A proinflammatory cytokine mixture consisting of IFN-γ, IL-1β and TNF-α highly increased CXCL11 mRNA expression and CXCL11 protein secretion by ARPE-19 cells. Resveratrol substantially inhibited the proinflammatory cytokines-induced CXCL11 production while partially blocking nuclear factor-κB activation. This inhibitory action of resveratrol was also observed for the cytokines-induced expression of chemokines CXCL9, CCL2 and CCL5. Our results indicate that resveratrol could potentially attenuate RPE inflammatory response implicated in the pathogenesis of AMD.
Sigma-Aldrich
Interferon-Inducible T Cell α Chemoattractant human, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Resveratrol, Vetec, reagent grade, 98%