Merck
  • IGFBP-2 inhibits adipogenesis and lipogenesis in human visceral, but not subcutaneous, adipocytes.
IGFBP-2 inhibits adipogenesis and lipogenesis in human visceral, but not subcutaneous, adipocytes.
International journal of obesity (2005) (2014-11-06)
S W Yau, V C Russo, I J Clarke, F R Dunshea, G A Werther, M A Sabin
ABSTRACT
IGF-binding protein (IGFBP)-2 is the principal IGFBP produced by white adipocytes during adipogenesis, and circulating levels are reduced in obesity. Overexpression of IGFBP-2 in transgenic mice prevents obesity, but depot-specific effects of IGFBP-2 on adipo/lipogenesis are unknown. The present study aimed to investigate whether IGFBP-2 affects adipo/lipogenesis in a depot-specific manner and explore potential mechanisms. Following adipocyte characterisation, IGFBP-2 levels were measured from human subcutaneous and visceral preadipocytes, and IGFBP-2 dose-responses were then undertaken with exogenous IGFBP-2 in an in vitro IGF-I-free system to examine adipo/lipogenesis. Following this, both types of adipocytes were transfected with human siRNA IGFBP-2 to assess auto-/para-/intra-crine effects, with and without additional add-back IGFBP-2. To elucidate the potential mechanisms, visceral preadipocytes were treated with either wild-type or Heparin Binding Domain (HBD)-mutant IGFBP-2 (which is unable to bind to cell-surface components), and experiments were also undertaken using Echistatin (an integrin receptor blocker). Outcomes included gene expression profiles, protein levels and phosphorylation and lipid staining. Human visceral adipocytes produced significantly more IGFBP-2 than subcutaneous adipocytes. Subsequent dose-responses to IGFBP-2 demonstrated significant reductions in adipo/lipogenesis in visceral, but not subcutaneous, adipocytes in response to increasing IGFBP-2. Silencing IGFBP-2 resulted in exaggerated adipo/lipogenesis in visceral, but not subcutaneous, adipocytes, an effect completely inhibited by add-back IGFBP-2. These effects occurred in the absence of changes in IGF-I levels. HBD-mutant IGFBP-2 had reduced effects compared with wild-type IGFBP-2. Wild-type IGFBP-2 increased phosphorylation of focal adhesion kinase (FAK) and decreased phosphatase and tensin homolog (PTEN) levels, suggestive of integrin-mediated signalling. Blockade of this signalling, using Echistatin, completely negated the effects of IGFBP-2 on visceral adipo/lipogenesis. IGFBP-2 inhibits both adipogenesis and lipogenesis in visceral, but not subcutaneous, adipocytes. This depot-specific impairment appears to be independent of IGF-I and involves cell-surface association of IGFBP-2 and activation of integrin signalling pathways.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human IGFBP2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Igfbp2