Skip to Content
Merck
All Photos(1)

Key Documents

D8893

Sigma-Aldrich

Dexamethasone

powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)

Synonym(s):

(11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-methylprednisolone, Prednisolone F

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$126.00

$126.00


Available to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 MG
$126.00

About This Item

Empirical Formula (Hill Notation):
C22H29FO5
CAS Number:
Molecular Weight:
392.46
Beilstein:
2066651
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77
biological source:
synthetic (organic)
form:
powder

$126.00


Available to ship on14 April 2025Details


Request a Bulk Order

biological source

synthetic (organic)

Quality Level

sterility

γ-irradiated

product line

BioXtra

Assay

≥80% (HPLC)

form

powder

potency

4-500 ng/mL

technique(s)

cell culture | mammalian: suitable

mp

262-264 °C (lit.)

solubility

ethanol: 1 mL/vial, clear

shipped in

ambient

storage temp.

2-8°C

SMILES string

C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO

InChI

1S/C22H29FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,17+,19+,20+,21+,22+/m1/s1

InChI key

UREBDLICKHMUKA-CXSFZGCWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dexamethasone has been used as a component of osteogenic differentiation medium.[1][2][3] It has also been used to study apoptosis, cell signaling pathways and gene expression.

Biochem/physiol Actions

Dexamethasone has immunosuppressant and antiemetic properties. It is used to treat postoperative nausea and vomiting (PONV) and chemotherapy-induced nausea and vomiting (CINV).[4] Dexamethasone plays an important role in cardiac transplant, sepsis, cancer and other pathologic disorders. It elevates the level of heat shock proteins.[5] It is an anti-inflammatory glucocorticoid with a range of effects on cell survival, cell signaling and gene expression.
Glucocorticoid anti-inflammatory agent
Glucocorticoid anti-inflammatory agent. Regulates T cell survival, growth, and differentiation. Inhibits the induction of nitric oxide synthase.

Physical form

powder 2-8 °C; stock-frozen in working aliquots, avoid repeated freeze/thaw

Reconstitution

To prepare 20 μg/ml stock solution: add 1 ml absolute ethanol; gently swirl to dissolve; add 49 ml sterile medium while mixing.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Unfolded Protein Response and Cellular Stress, Part 2 (2011)
Substrate elasticity regulates adipose-derived stromal cell differentiation towards osteogenesis and adipogenesis through $\beta$-catenin transduction
Xie J, et al.
Acta Biomaterialia, 79(5), 83-95 (2018)
Bimolecular based heparin and self-assembling hydrogel for tissue engineering applications
Fernandez-Muinos T, et al.
Acta Biomaterialia, 16(5), 35-48 (2015)
Pharmacology and Physiology for Anesthesia: Foundations and Clinical Application (2013)
C A Smith et al.
Journal of tissue engineering and regenerative medicine, 9(5), 595-604 (2014-06-20)
Fresh-frozen biological allograft remains the most effective substitute for the 'gold standard' autograft, sharing many of its osteogenic properties but, conversely, lacking viable osteogenic cells. Tissue engineering offers the opportunity to improve the osseointegration of this material through the addition

Questions

1–6 of 6 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the solubility of Product D8893, Dexamethasone, in ethanol?

    1 answer
    1. The product is soluble in 95% ethanol (1mg/mL), yielding a clear, colorless solution. A 20 μg/mL stock solution can be formed by adding 49 mL of media to 1 mL of a 1 mg/mL ethanol stock.

      Helpful?

  3. What is the best way to dissolve Product D8893, Dexamethasone?

    1 answer
    1. The solubility of dexamethasone is <0.01 mg/mL in water and 24 mg/mL in H5784 (a 45% w/v solution of 2-hydroxypropyl-β-cyclodextrin).

      Helpful?

  4. How do the dexamethasone products D8893 and the D4902 compare?

    1 answer
    1. They are the same except for the sterility. D8893 has been g-irradiated, so it is sterile. The D4902 is not necessarily sterile. Both are cell culture tested by Sigma.

      Helpful?

  5. What is the solubility of Product D8893, Dexamethasone, in DMSO?

    1 answer
    1. The solubility of dexamethasone in DMSO is 606 mg/mL at 25°C.

      Helpful?

  6. What solution stability information do you have available for Product D8893, Dexamethasone?

    1 answer
    1. We would recommend that you store stock solutions in working aliquots frozen at -20°C. It is best to avoid repeated freeze/thaw cycles.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service