Merck
  • Physiological and proteome analysis suggest critical roles for the photosynthetic system for high water-use efficiency under drought stress in Malus.
Physiological and proteome analysis suggest critical roles for the photosynthetic system for high water-use efficiency under drought stress in Malus.
Plant science : an international journal of experimental plant biology (2015-05-31)
Shasha Zhou, Mingjun Li, Qingmei Guan, Fengli Liu, Sheng Zhang, Wei Chen, Lihua Yin, Yuan Qin, Fengwang Ma
ABSTRACT
Water use efficiency is an important indicator for plant adaptation and resistance to drought conditions. We previously found that under moderate drought stress, the water use efficiency of cv. 'Qinguan' apple (Malus domestica Borkh.) (tolerant to drought) was enhanced, while that of cv. 'Naganofuji No. 2' was not enhanced. In this research, we also found that instantaneous water-use efficiency of cv. 'Qinguan' was higher than that of cv. 'Naganofuji No. 2', mainly because of its higher net photosynthesis rate. To dissect the potential mechanisms underlying this phenomenon, we performed a comparative iTRAQ-based proteomics analysis with leaves of drought-treated cv. 'Qinguan' and 'Naganofuji No. 2'. We identified 4078 proteins, of which 594 were differentially abundant between drought and well-watered leaves. The majority of increased proteins were predicted to be involved in photosynthetic pathway in drought treated cv. 'Qinguan' leaves, indicating that regulation of photosynthesis plays an important role for higher water use efficiency under drought stress. Enzyme activity assays were performed to validate the proteomics data. Our results suggested that the main regulatory mechanisms for high water use efficiency of cv. 'Qinguan' under moderate drought stress included the maintaining of Calvin cycle function by increasing key enzymes, stabilization of photosynthetic electron transfer and keeping reactive oxygen species at normal level by regulation of photosynthetic electron transfer chain, photorespiration and reactive oxygen species scavenging capability, thus prevented photoinhibition, reduced reactive oxygen species production and enhanced net photosynthesis rate. In addition, the response of signal regulatory proteins and abiotic stress-responsive proteins to drought also helped plants to cope with such stress.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SAFC
Glycine
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)
Sigma-Aldrich
S-Methyl methanethiosulfonate, 97%