Merck
  • Multiresidue analysis of 36 pesticides in soil using a modified quick, easy, cheap, effective, rugged, and safe method by liquid chromatography with tandem quadruple linear ion trap mass spectrometry.
Multiresidue analysis of 36 pesticides in soil using a modified quick, easy, cheap, effective, rugged, and safe method by liquid chromatography with tandem quadruple linear ion trap mass spectrometry.
Journal of separation science (2015-06-23)
Xue Feng, Zeying He, Lu Wang, Yi Peng, Ming Luo, Xiaowei Liu
ABSTRACT
A new method for simultaneous determination of 36 pesticides, including 15 organophosphorus, six carbamate, and some other pesticides in soil was developed by liquid chromatography with tandem quadruple linear ion trap mass spectrometry. The extraction and clean-up steps were optimized based on the quick, easy, cheap, effective, rugged, and safe method. The data were acquired in multiple reaction monitoring mode combined with enhanced product ion to increase confidence of the analytical results. Validation experiments were performed in soil samples. The average recoveries of pesticides at four spiking levels (1, 5, 50, and 100 μg/kg) ranged from 63 to 126% with relative standard deviation below 20%. The limits of detection of pesticides were 0.04-0.8 μg/kg, and the limits of quantification were 0.1-2.6 μg/kg. The correlation coefficients (r(2) ) were higher than 0.990 in the linearity range of 0.5-200 μg/L for most of the pesticides. The method allowed for the analysis of the target pesticides in the lower μg/kg concentration range. The optimized method was then applied to the test of real soil samples obtained from several areas in China, confirming the feasibility of the method.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Profenofos, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture