Merck
  • Acute inhibition of monoamine oxidase and ischemic preconditioning in isolated rat hearts: interference with postischemic functional recovery but no effect on infarct size reduction.
Acute inhibition of monoamine oxidase and ischemic preconditioning in isolated rat hearts: interference with postischemic functional recovery but no effect on infarct size reduction.
Canadian journal of physiology and pharmacology (2015-09-01)
Maria D Dănilă, Andreea I Privistirescu, Silvia N Mirica, Adrian Sturza, Valentin Ordodi, Lavinia Noveanu, Oana M Duicu, Danina M Muntean
ABSTRACT
Monoamine oxidases (MAOs) have recently emerged as important mitochondrial sources of oxidative stress in the cardiovascular system. Generation of reactive oxygen species during the brief episodes of ischemic preconditioning (IPC) is responsible for the cardioprotection at reperfusion. The aim of this study was to assess the effects of two MAO inhibitors (clorgyline and pargyline) on the IPC-related protection in isolated rat hearts. Animals subjected to 30 min global ischemia and 120 min reperfusion were assigned to the following groups: (i) Control, no additional intervention; (ii) IPC, 3 cycles of 5 min ischemia and 5 min reperfusion before the index ischemia; (iii) IPC-clorgyline, IPC protocol bracketed for 5 min with clorgyline (50 μmol/L); (iv) IPC-pargyline, IPC protocol bracketed for 5 min with pargyline (0.5 mmol/L). The postischemic functional recovery was assessed by the left ventricular developed pressure (LVDP) and the indices of contractility (+dLVP/dt max) and relaxation (-dLVP/dt max). Infarct size (IS) was quantified by TTC staining. In both genders, IPC significantly improved functional recovery that was further enhanced in the presence of either clorgyline or pargyline. IS reduction was comparable among all the preconditioned groups, regardless of the presence of MAO inhibitors. In isolated rat hearts, acute inhibition of MAOs potentiates the IPC-induced postischemic functional recovery without interfering with the anti-necrotic protection.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, Vetec, reagent grade, 99%