Merck
  • Synthesis, luminescence, and electrochemical studies of a new series of octanuclear ruthenium(II) complexes of tolylterpyridine appended calixresorcarenes.
Synthesis, luminescence, and electrochemical studies of a new series of octanuclear ruthenium(II) complexes of tolylterpyridine appended calixresorcarenes.
Dalton transactions (Cambridge, England : 2003) (2015-07-30)
Selvam Amudhan Senthan, Vedamanickam Alexander
ABSTRACT
A new series of octanuclear Ru(ii) complexes of tolylterpyridine appended calixresorcarenes [{Ru(ttpy)}8()](PF6)16 (), [{Ru(ttpy)}8()](PF6)16 (), and [{Ru(ttpy)}8()](PF6)16 () [ = 2,8,14,20-tetraethyl-4,6,10,12,16,18,22,24-octa(4'-p-benzyloxy-(2,2':6',2''-terpyridinyl))calix[4]resorcarene; = 2,8,14,20-tetraphenyl-4,6,10,12,16,18,22,24-octa(4'-p-benzyloxy-(2,2':6',2''-terpyridinyl))calix[4]resorcarene; and = 2,8,14,20-tetra-p-tolyl-4,6,10,12,16,18,22,24-octa(4'-p-benzyloxy-(2,2':6',2''-terpyridinyl))calix[4]resorcarene] have been synthesized and characterized. The tetraethyl-, tetraphenyl-, and tetra-p-tolylcalixresorcarenes (), (), and (), respectively, are characterized by single crystal X-ray diffraction: () a = 13.5771(4) Å, b = 7.9566(3) Å, c = 18.3912(6) Å, α = γ = 90°, β = 103.146(2)°, V = 1934.69(11) Å(3), Z = 2, monoclinic, P2/n; () a = 11.9710(8) Å, b = 12.7057(9) Å, c = 13.8570(9) Å, α = 94.731(2)°, β = 108.558(2)°, γ = 106.657(2)°, V = 1878.9(2) Å(3), Z = 1, triclinic, P1[combining macron]; and () a = 26.076(3) Å, b = 20.056(2) Å, c = 16.0193(19) Å, α = γ = 90°, β = 116.056(3)°, V = 7526.3(14) Å(3), Z = 4, monoclinic, C2/c, respectively. The octanuclear complexes are nonluminescent at 298 K, but exhibit (3)MLCT luminescence at 654, 649, and 646 nm, respectively, at 77 K in acetonitrile. In the solid state at 298 K they exhibit (3)MLCT luminescence at 658, 658, and 665 nm, respectively. The complexes exhibit a monoexponential decay profile in acetonitrile and in the solid state at 298 K. The redox active hosts containing eight [Ru(ttpy)2](2+) moieties undergo quasireversible one-electron oxidation at 1.25, 1.26, and 1.25 V versus Ag/Ag(+), respectively, in acetonitrile. The study demonstrates the versatility of the octanucleating calixresorcarene based ligands , , and in forming octanuclear Ru(ii) complexes.
Sigma-Aldrich
Potassium hexafluorophosphate, 99.5% trace metals basis
Sigma-Aldrich
2-Acetylpyridine, ≥99%, FG
Sigma-Aldrich
Resorcinol, meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Resorcinol, ≥98%, FG
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Potassium hexafluorophosphate, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
2-Acetylpyridine, ≥99%
Sigma-Aldrich
p-Tolualdehyde, 97%
Sigma-Aldrich
Propionaldehyde, ≥97%, FG
Sigma-Aldrich
Cesium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Cesium carbonate, 99.9% trace metals basis
Sigma-Aldrich
p-Tolualdehyde, ≥97%, FG
Sigma-Aldrich
Cesium carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Cesium carbonate, 99.95% trace metals basis
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Resorcinol, BioXtra, ≥99%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%