Merck
  • Validation of an HPLC-MS/MS and wipe procedure for mitomycin C contamination.
Validation of an HPLC-MS/MS and wipe procedure for mitomycin C contamination.
Journal of chromatographic science (2014-08-19)
Clayton B'Hymer, Thomas Connor, Derek Stinson, Jack Pretty
ABSTRACT
A high-performance liquid chromatography-tandem mass spectrometric (HPLC-MS/MS) method was developed for the determination of mitomycin C, an anticancer drug, from contamination on various surfaces. Mitomycin C is often used in various forms of intraperitoneal chemotherapy, and operating room healthcare worker exposure to this drug is possible. The surface testing method consisted of a wiping procedure utilizing a solution of 20/45/35 (v/v/v) of acetonitrile-isopropanol-water made 0.01 M in ammonium citrate (apparent pH 7.0). The wipe solutions were analyzed by means of HPLC-MS/MS using a reversed-phase gradient system and electrospray ionization in positive ion mode with a triple-quadrupole MS detector. Accuracy and precision of this method were demonstrated by a series of recovery studies of both spiked solutions and extracted wipes from various surfaces (stainless steel, vinyl and Formica(®)) spiked with known levels of mitomycin C. Recoveries of spiked solutions containing the analyte demonstrate mean recoveries (accuracy) ranged from 93 to 105%. Precision as measured by the relative standard deviation (% RSD) of multiple samples (n= 10) at each concentration level demonstrated values of 7.5% or less. The recoveries from spiked surfaces varied from 30 to 99%. The limit of detection for this methodology is ∼2 ng/100 cm(2) equivalent surface area, and the limit of quantitation is ∼6 ng/100 cm(2).
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Ammonium citrate tribasic, ≥97% (titration)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Citric acid monohydrate, BioXtra, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Citric acid monohydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, E330, 99.5-100.5% (based on anhydrous substance), grit
Sigma-Aldrich
Citric acid monohydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Citric acid monohydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, 99.5-102%
Sigma-Aldrich
Citric acid monohydrate, reagent grade, ≥98% (GC/titration)
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%