Merck
  • Human urinary renalase lacks the N-terminal signal peptide crucial for accommodation of its FAD cofactor.
Human urinary renalase lacks the N-terminal signal peptide crucial for accommodation of its FAD cofactor.
International journal of biological macromolecules (2015-04-26)
Valerii I Fedchenko, Olga A Buneeva, Arthur T Kopylov, Alexander V Veselovsky, Victor G Zgoda, Alexei E Medvedev
ABSTRACT
Renalase is a recently discovered secretory protein involved in the regulation of blood pressure. Cells synthesize all known isoforms of human renalase (1 and 2) as flavoproteins. Accommodation of FAD in the renalase protein requires the presence of its N-terminal peptide. However, in secretory proteins, such peptides are usually cleaved during their export from the cell. In the present study, we have isolated human renalase from urinary samples of healthy volunteers and human recombinant renalases 1 and 2 expressed in Escherichia coli cells. In these proteins, we investigated the presence of the renalase N-terminal peptide and the FAD cofactor and performed computer-aided molecular analysis of the renalase crystal structure to evaluate possible consequences of removal of the N-terminal peptide. In contrast to human recombinant renalase isoforms 1 and 2 containing non-covalently bound FAD and clearly detectable N-terminal peptide, renalase purified from human urine lacks both the N-terminal signal peptide and FAD. The computer-aided analysis indicates that the removal of this peptide results in inability of the truncated renalase to bind the FAD cofactor. Thus, our results indicate that human renalase secreted in urine lacks its N-terminal peptide, and therefore catalytic activities of urinary renalase reported in the literature cannot be attributed to FAD-dependent mechanisms. We suggest that FAD-dependent catalytic functions are intrinsic properties of intracellular renalases, whereas extracellular renalases act in FAD- and possibly catalytic-independent manner.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Heptafluorobutyric acid, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Heptafluorobutyric acid, 98%
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)