Merck
  • Validation of a streamlined multiclass, multiresidue method for determination of veterinary drug residues in bovine muscle by liquid chromatography-tandem mass spectrometry.
Validation of a streamlined multiclass, multiresidue method for determination of veterinary drug residues in bovine muscle by liquid chromatography-tandem mass spectrometry.
Analytical and bioanalytical chemistry (2014-12-30)
Marilyn J Schneider, Steven J Lehotay, Alan R Lightfield
ABSTRACT
Multiclass, multiresidue methods are becoming increasingly popular in regulatory monitoring programs due to their increased analytical scope and laboratory efficiency. In this work, we report the development and validation of a new high-throughput analytical method to monitor up to 131 veterinary drug residues, representing at least 13 different classes, in bovine muscle. This novel method streamlined sample preparation to <15 min/sample/analyst, or a batch of 40-60 pre-homogenized samples in <3 h/analyst, through the combination of dispersive solid-phase extraction with in-vial filtration (a new technique known as filter-vial d-SPE). The use of an enhanced sensitivity state-of-the-art tandem mass spectrometer led to <10 ng/g limits of quantification for nearly all drug analytes with injection of 0.17 mg of equivalent sample. Positive and negative switching in electrospray ionization was applied to cover all analytes in an 11-min liquid chromatographic separation. In the 3-day validation study, 100 of the drugs met quantification criteria of 70-120% recoveries and Horwitz Ratio ≤1.0, and the remaining analytes could still be screened at regulatory target levels. In the validation study involving >11,400 analyte results for spiked samples, the rate of false negatives for identification purposes was <5%, and no false positives occurred at appreciable concentrations.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Thiamphenicol
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Sigma-Aldrich
2-Thiouracil, ≥99%
Sigma-Aldrich
Metronidazole, BioXtra
Sigma-Aldrich
2-Quinoxalinecarboxylic acid, 97%
Sigma-Aldrich
2-Mercapto-1-methylimidazole, ≥99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Tetracycline hydrochloride, powder
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracycline hydrochloride, meets USP testing specifications
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
6-Phenyl-2-thiouracil, ≥95%
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Niclosamide
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis