Merck
  • Quantitative determination of isoquinoline alkaloids and chlorogenic acid in Berberis species using ultra high performance liquid chromatography with hybrid triple quadrupole linear ion trap mass spectrometry.
Quantitative determination of isoquinoline alkaloids and chlorogenic acid in Berberis species using ultra high performance liquid chromatography with hybrid triple quadrupole linear ion trap mass spectrometry.
Journal of separation science (2015-04-08)
Awantika Singh, Vikas Bajpai, Sunil Kumar, Kamal Ram Arya, Kulwant Rai Sharma, Brijesh Kumar
ABSTRACT
Berberis species are well known and used extensively as medicinal plants in traditional medicine. They have many medicinal values attributable to the presence of alkaloids having different pharmacological activities. In this study, a method was developed and validated as per international conference on harmonization guidelines using ultra high performance liquid chromatography with hybrid triple quadrupole-linear ion trap mass spectrometry operated in the multiple reaction monitoring mode for nine bioactive compounds, including protoberberine alkaloids, aporphine alkaloids and chlorogenic acid. This method was applied in different plant parts of eight Berberis species to determine variations in content of nine bioactive compounds. The separation was achieved on an ACQUITY UPLC CSH™ C18 column using a gradient mobile phase at flow rate 0.3 mL/min. Calibration curves for all the nine analytes provided optimum linear detector response (with R(2) ≥0.9989) over the concentration range of 0.5-1000 ng/mL. The precision and accuracy were within RSDs ≤2.4 and ≤2.3%, respectively. The results indicated significant variation in the total contents of the nine compounds in Berberis species.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis