Merck
  • Detection and characterization of betamethasone metabolites in human urine by LC-MS/MS.
Detection and characterization of betamethasone metabolites in human urine by LC-MS/MS.
Drug testing and analysis (2014-12-30)
Xavier Matabosch, Oscar J Pozo, Núria Monfort, Clara Pérez-Mañá, Magí Farré, Jordi Segura, Rosa Ventura
ABSTRACT
Glucocorticosteroids are prohibited in sports when administered by systemic routes and allowed using other administrations for therapeutic reasons. Therefore, markers to distinguish between routes of administration through the analysis of urine samples are needed in anti-doping control. As a first step to achieve that goal, the metabolism of betamethasone (BET) was investigated in the present work. Urine samples obtained after BET intramuscular injection were hydrolyzed with β-glucuronidase and subjected to liquid-liquid extraction with ethyl acetate in alkaline conditions. The extracts were analyzed by liquid chromatography coupled to tandem mass spectrometry. Common open screening methods for fluorine containing corticosteroids (precursor ion scan method of m/z 121, 147, 171, and neutral loss (NL) scan methods of 20 and 38 Da in positive ionization, and 46 and 76 Da in negative ionization) were applied to detect BET metabolites. Moreover, an NL method was applied to detect A-ring reduced metabolites of BET, which are ionized as [M+NH4 ](+) (NL of 55, 73, and 91 Da, corresponding to the consecutive losses of NH3 , HF and one, two and three water molecules, respectively). BET and 24 metabolites were detected. Six metabolites were identified by comparison with standards, and for ten, feasible structures were proposed based on mass spectrometric data. Eleven of the characterized metabolites had not been previously reported. Metabolites resulting from 11-oxidation, 6-hydroxylation, C20 or 4-ene-3-one reduction and combination of some of them were detected. Moreover one metabolite resulting from cleavage of the side chain with subsequent oxidation of carbon at C17 was also detected.
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri