Merck
  • Mycotoxins in Plant-Based Dietary Supplements: Hidden Health Risk for Consumers.
Mycotoxins in Plant-Based Dietary Supplements: Hidden Health Risk for Consumers.
Journal of agricultural and food chemistry (2015-07-15)
Zdenka Veprikova, Milena Zachariasova, Zbynek Dzuman, Alena Zachariasova, Marie Fenclova, Petra Slavikova, Marta Vaclavikova, Katerina Mastovska, Daniel Hengst, Jana Hajslova
ABSTRACT
Mycotoxin contamination of dietary supplements represents a possible risk for human health, especially in the case of products intended for people suffering from certain health conditions. The aim of this study was to assess the extent of this problem based on analyses of a wide set of herbal-based dietary supplements intended for various purposes: (i) treatment of liver diseases (milk thistle); (ii) reduction of menopause effects (red clover, flax seed, and soy); and (iii) preparations for general health support (green barley, nettle, goji berries, yucca, etc.) The analytical method including 57 mycotoxins was based on a QuEChERS-like (quick, easy, cheap, effective, rugged, safe) approach and ultrahigh performance liquid chromatography coupled with tandem mass spectrometry. The main mycotoxins determined were Fusarium trichothecenes, zearalenone and enniatins, and Alternaria mycotoxins. Co-occurrence of enniatins, HT-2/T-2 toxins, and Alternaria toxins was observed in many cases. The highest mycotoxin concentrations were found in milk thistle-based supplements (up to 37 mg/kg in the sum).
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Sigma-Aldrich
Dimethyl ether, ≥99%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Verrucarol
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Ammonium acetate, Vetec, reagent grade, 97%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet