Merck
  • Quantification of neurosteroids during pregnancy using selective ion monitoring mass spectrometry.
Quantification of neurosteroids during pregnancy using selective ion monitoring mass spectrometry.
Steroids (2014-12-30)
Kurt D Pennell, Mark A Woodin, Page B Pennell
ABSTRACT
Analytical techniques used to quantify neurosteroids in biological samples are often compromised by non-specificity and limited dynamic range which can result in erroneous results. A relatively rapid and inexpensive gas chromatography-mass spectrometry (GC-MS) was developed to simultaneously measure nine neurosteroids, including allopregnanolone, estradiol, and progesterone, as well as 25-hydroxy-vitamin D3 in plasma samples collected from adult women subjects during and after pregnancy. Sample preparation involved solid-phase extraction and derivatization, followed by automated injection on a GC equipped with a mass selective detector (MSD) operated in single ion monitoring (SIM) mode to yield a run time of less than 11min. Method detection limits for all neurosteroids ranged from 30 to 200pg/mL (parts per trillion), with coefficients of variation that ranged from 3% to 5% based on intra-assay comparisons run in triplicate. Although concentrations of estradiol measured by chemiluminescent immunoassay (CIA) were consistent with values determined by GC-MS values, CIA yielded considerable higher values of progesterone, suggesting antibody cross reactions resulting from low specificity. Mean neurosteroid levels and representative time-course data demonstrate the ability of the method to quantify changes in multiple neurosteroids during pregnancy, including rapid declines in neurosteroid levels associated with delivery. This simplified GC-MS method holds particular promise for research and clinical laboratories that require simultaneous quantification of multiple neurosteroids, but lack the resources and expertise to support advanced liquid chromatography-tandem mass spectrometry facilities.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
25-Hydroxyvitamin D3 monohydrate, ≥99.0% (HPLC)
Supelco
Methanol, analytical standard
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 1% TMCS, 99% (excluding TMCS)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%
Sigma-Aldrich
5α-Pregnan-3α-ol-20-one, solid
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Sigma-Aldrich
5β-Pregnan-3α-ol-20-one
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Estrone, ≥99%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Supelco
Progesterone, VETRANAL®, analytical standard
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 10% TMCS, 98% (excluding TMCS)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Estrone, European Pharmacopoeia (EP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Supelco
Estrone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material