Merck
  • An improved liquid chromatography-tandem mass spectrometric method to quantify formoterol in human urine.
An improved liquid chromatography-tandem mass spectrometric method to quantify formoterol in human urine.
Journal of chromatographic science (2013-09-14)
Genye He, Jianghai Lu, Xiaobing Wang, Youxuan Xu, Yun Wu, Ying Dong, Li Shen, Zhenwen He, Jian Zhao, Hong Yuan
ABSTRACT
Formoterol is a new threshold substance in the prohibited list 2012 according to World Anti-Doping Agency. Extracted by ethyl acetate using formoterol-D6 as internal standard, formoterol underwent a constant flow rate gradient elution separation in reversed-phase liquid chromatography. Subsequently, mass spectrometry analysis was conducted by tandem mass spectrometry in the multiple reaction monitoring mode. According to the proposed method, a calibration curve was constructed in the range of 0.2-500 ng/mL (r(2) = 1) with a limit of quantification 0.2 ng/mL. The mean extracted recovery assessed at three different concentrations (1, 30 and 100 ng/mL) was more than 80%. The method was validated by the analysis of three quality control samples from World Association of Anti-Doping Scientists. In conclusion, the developed and validated method was sensitive, accurate and precise for the quantification of formoterol in human urine for doping control purposes.
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, BioReagent, suitable for electrophoresis, 98.0-102.0%
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, BioXtra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, puriss. p.a., ACS reagent, ≥99.0% (T)
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, ACS reagent, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%