Merck
  • Enhancing perception of contaminated food through acid-mediated modulation of taste neuron responses.
Enhancing perception of contaminated food through acid-mediated modulation of taste neuron responses.
Current biology : CB (2014-08-19)
Yan Chen, Hubert Amrein
ABSTRACT
Natural foods contain not only nutrients, but also nonnutritious and potentially harmful chemicals. Thus, animals need to evaluate food content in order to make adequate feeding decisions. Here, we investigate the effects of acids on the taste neuron responses and on taste behavior of desirable, nutritious sugars and sugar/bitter compound mixtures in Drosophila melanogaster. Using Ca2+ imaging, we show that acids activate neither sweet nor bitter taste neurons in tarsal taste sensilla. However, they suppress responses to bitter compounds in bitter-sensing neurons. Moreover, acids reverse suppression of bitter compounds exerted on sweet-sensing neurons. Consistent with these observations, behavioral analyses show that bitter-compound-mediated inhibition on feeding behavior is alleviated by acids. To investigate the cellular mechanism by which acids modulate these effects, we silenced bitter-sensing gustatory neurons. Surprisingly, this intervention had little effect on acid-mediated derepression of sweet neuron or feeding responses to either sugar/bitter compound mixtures or sugar/bitter compound/acid mixtures, suggesting that there are two independent pathways by which bitter compounds are sensed. Our investigations reveal that acids, when presented in dietary relevant concentrations, enhance the perception of sugar/bitter compound mixtures. Drosophila's natural food sources-fruits and cohabitating yeast-are rich in sugars and acids but are rapidly colonized by microorganisms, such as fungi, protozoan parasites, and bacteria, many of which produce bitter compounds. We propose that the acids present in most fruits counteract the inhibitory effects of these bitter compounds during feeding.
Product Description
Supelco
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
Denatonium benzoate, ≥98%
Sigma-Aldrich
D-(−)-Fructose, 98.0-102.0% dry basis, meets USP testing specifications
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Acetic acid, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
D-(−)-Fructose, tested according to Ph. Eur.
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Millipore
D-(−)-Fructose, ≥99.0% (HPLC), suitable for microbiology
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Fructose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Fructose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC