Merck
  • Quantification of furanic derivatives in fortified wines by a highly sensitive and ultrafast analytical strategy based on digitally controlled microextraction by packed sorbent combined with ultrahigh pressure liquid chromatography.
Quantification of furanic derivatives in fortified wines by a highly sensitive and ultrafast analytical strategy based on digitally controlled microextraction by packed sorbent combined with ultrahigh pressure liquid chromatography.
Journal of chromatography. A (2015-01-27)
Rosa Perestrelo, Catarina L Silva, José S Câmara
ABSTRACT
An improved, reliable and powerful analytical strategy based on digitally controlled microextraction by packed sorbent (MEPS) combined with ultrahigh pressure liquid chromatography (UHPLC) was validated for the simultaneous identification and quantification of major furanic derivatives, namely 5-hydroxymethyl-2-furaldehyde (5HMF), 5-methyl-2-furaldehyde (5MF), 2-furaldehyde (2F) and 2-furyl methyl ketone (2FMK), in fortified wines. To enhance the extraction efficiency of the target furanic derivates, several influencing extraction parameters, such as number of loading cycles, nature of elution solvent and elution volume, were evaluated and optimized. In addition the ability of different MEPS sorbent materials, namely C2, C8, C18, SIL, M1, R-AX, R-CX and PGC, were also tested. The optimal analytical conditions involved loading 3×200 μL of wine samples through a C8 sorbent in a MEPS syringe placed in the semi-automatic eVolH syringe followed by elution using 200 μL MeOH:H2O (95:5, v/v). The furanic derivates separation was achieved using a CORTECS UPLC(®) C18 analytical column in an ultrafast chromatographic run (within 4 min). The method performance was assessed for dry/medium dry (D/MD) and sweet/medium sweet (S/MS) model wines in terms of selectivity, linearity, limit of detection (LOD), limit of quantitation (LOQ), accuracy, precision and matrix effect, using model wine matrix-matched calibration. Good linearity was obtained with a regression coefficient (r(2)) higher than 0.992. A good precision was attained (RSD<5%) and low LODs were achieved for D/MD (4.5-129.3 ng L(-1)) and S/MS (6.9-285.2 ng L(-1)) model wines. The quantification limits (LOQ) for D/MD model wines ranged from 14.9 to 431.0 ng L(-1), whereas for S/MS model wines range from 23.1 to 950.5 ng L(-1). The method also afforded satisfactory results in terms of accuracy, ranging from 74 to 97% for D/MD wines and between 84 and 99% for S/MS wines. The MEPS(C8)/UHPLC-PDA analytical strategy was successfully applied to analyze furanic derivates in 26 fortified Madeira wines from different types (D/MD, S/MS) and ages. The obtained results revealed the analytical strategy as a suitable tool which combines sensitivity, effectiveness, reduced analysis time and simple analytical procedure. Principal component analysis (PCA) suggested that fortified wines can be organized based on their age on PC1, which are mainly characterized by 5HMF.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, for use with autosampler
Sigma-Aldrich
2-Furyl methyl ketone, natural (US), ≥97%, FG
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with manual holder
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with manual holder
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with manual holder, needle size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with manual holder
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
2-Furyl methyl ketone, 99%
Sigma-Aldrich
Furfural, ≥98%, FCC, FG