Merck
  • Discovery of novel small-molecule compounds with selective cytotoxicity for Burkitt's lymphoma cells using 3D ligand-based virtual screening.
Discovery of novel small-molecule compounds with selective cytotoxicity for Burkitt's lymphoma cells using 3D ligand-based virtual screening.
Molecules (Basel, Switzerland) (2014-11-22)
Martina Gobec, Izidor Sosič, Boris Brus, Aleš Obreza, Stanislav Gobec, Irena Mlinarič-Raščan
ABSTRACT
We describe a ligand-based approach towards compounds with more specific targeting for Burkitt's lymphoma. Using three-dimensional ligand-based similarity searches and a previously described hit compound, we have identified six compounds that are chemically different but with similar spatial conformations. Biological evaluation revealed that one compound has better growth inhibition and improved selectivity towards Burkitt's lymphoma cells than the query compound. However, initial mechanism-of-action studies show a different target profile in comparison with the previous hit compound, which does not involve the inhibition of the proteasome or the NFκB pathway. The data from this study provide a solid basis for further efforts in the search for selective agents against Burkitt's lymphoma.
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Supelco
Water, for TOC analysis
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Water, suitable for ion chromatography
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality