Skip to Content
Merck
All Photos(3)

Key Documents

185965

Sigma-Aldrich

Dimethyl sulfoxide-d6

99.9 atom % D, contains 1 % (v/v) TMS

Synonym(s):

(Methyl sulfoxide)-d6, DMSO-d6, Hexadeuterodimethyl sulfoxide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$72.30
25 G
$170.00
50 G
$327.00

$72.30


Available to ship on24 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
$72.30
25 G
$170.00
50 G
$327.00

About This Item

Linear Formula:
(CD3)2SO
CAS Number:
Molecular Weight:
84.17
Beilstein:
1237248
MDL number:
UNSPSC Code:
12191502
eCl@ss:
39092001
PubChem Substance ID:
NACRES:
NA.21

$72.30


Available to ship on24 April 2025Details


Request a Bulk Order

vapor pressure

0.42 mmHg ( 20 °C)

Quality Level

isotopic purity

99.9 atom % D

Assay

≥99% (CP)

form

liquid

autoignition temp.

573 °F

contains

1 % (v/v) TMS

expl. lim.

42 %

technique(s)

NMR: suitable

impurities

≤0.0250% water
water

refractive index

n20/D 1.476 (lit.)

bp

189 °C (lit.)

mp

20.2 °C (lit.)

density

1.190 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3/i1D3,2D3

InChI key

IAZDPXIOMUYVGZ-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sulfoxide-d6 (DMSO-d6) is a deuterated solvent[1] and it contains 1%(v/v) TMS (Tetramethylsilane, added as a stabilizer). On photoirradiation in the range of 193 and 222nm, it undergoes decomposition to afford CD3 radicals. Quantum yields of CD3 have been evaluated by infrared diode laserabsorption spectroscopy.[2] 100% DMSO-d6 has been used as solvent in the long-range COSY (Correlation Spectroscopy) experiment.[3]

Application

DMSO-d6 along with glycerol or glycerol-d8 forms highly viscous binary solvents with high dissolution power for complex biological active compounds.[4]

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly Viscous Binary Solvents: DMSO-d6/Glycerol and DMSO-d6/Glycerol-d8 for Polar and Apolar Mixture Analysis by NMR.
Lameiras P and Nuzillard J-M.
AZo Journal of Materials Online, 88(8), 4508-4515 (2016)
M G Nair et al.
Applied and environmental microbiology, 57(2), 434-439 (1991-02-01)
Two isoflavonoids isolated from clover roots grown under phosphate stress were characterized as formononetin (7-hydroxy,4'-methoxy isoflavone) and biochanin A (5,7-dihydroxy,4'-methoxy isoflavone). At 5 ppm, these compounds stimulated hyphal growth in vitro and root colonization of an undescribed vesicular-arbuscular mycorrhiza, a
Using high-performance quantitative NMR (HP-qNMR?) for certifying traceable and highly accurate purity values of organic reference materials with uncertainties< 0.1%.
Weber M, et al.
Accreditation and Quality Assurance, 18(2), 91-98 (2013)
Diode laser measurements of CD3 quantum yields and internal energy for the dissociation of dimethyl sulfoxide-d6.
Rudolph RN, et al.
J. Chem. Phys. , 106(4), 1346-1352 (1997)
Papawee Suabjakyong et al.
PloS one, 10(3), e0122733-e0122733 (2015-03-27)
The basidiomycetous mushroom Phellinus igniarius (L.) Quel. has been used as traditional medicine in various Asian countries for many years. Although many reports exist on its anti-oxidative and anti-inflammatory activities and therapeutic effects against various diseases, our current knowledge of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service