Merck
  • Histopathological and biomechanical evaluation of tenocyte seeded allografts on rat Achilles tendon regeneration.
Histopathological and biomechanical evaluation of tenocyte seeded allografts on rat Achilles tendon regeneration.
Biomaterials (2015-03-17)
Cansın Güngörmüş, Dürdane Kolankaya, Erkin Aydin
ABSTRACT
Tendon injuries in humans as well as in animals' veterinary medicine are problematic because tendon has poor regenerative capacity and complete regeneration of the ruptured tendon is never achieved. In the last decade there has been an increasing need of treatment methods with different approaches. The aim of the current study was to improve the regeneration process of rat Achilles tendon with tenocyte seeded decellularized tendon matrices. For this purpose, Achilles tendons were harvested, decellularized and seeded as a mixture of three consecutive passages of tenocytes at a density of 1 × 10(6) cells/ml. Specifically, cells with different passage numbers were compared with respect to growth characteristics, cellular senescence and collagen/tenocyte marker production before seeding process. The viability of reseeded tendon constructs was followed postoperatively up to 6 months in rat Achilles tendon by histopathological and biomechanical analysis. Our results suggests that tenocyte seeded decellularized tendon matrix can significantly improve the histological and biomechanical properties of tendon repair tissue without causing adverse immune reactions. To the best of our knowledge, this is the first long-term study in the literature which was accomplished to prove the use of decellularized matrix in a clinically relevant model of rat Achilles tendon and the method suggested herein might have important implications for translation into the clinic.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Triton X-100, laboratory grade
Supelco
Dichloromethane, analytical standard
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer